Product Details

SNP ID
rs11550918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65086494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCACCTGCCGGCTATCAGTGCCC[C/T]AGCTGCAATGGCCCCATCTTCCCCC
Phenotype
MIM: 609374 MIM: 615738
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDCA5 PubMed Links

Gene Details

Gene
CDCA5
Gene Name
cell division cycle associated 5
There are no transcripts associated with this gene.

Gene
TMEM262
Gene Name
transmembrane protein 262
There are no transcripts associated with this gene.

Gene
VPS51
Gene Name
VPS51, GARP complex subunit
There are no transcripts associated with this gene.

Gene
ZFPL1
Gene Name
zinc finger protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006782.3 459 Silent Mutation CCC,CCT P98P NP_006773.2

View Full Product Details