Product Details

SNP ID
rs7124215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65170595 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGAGGAGCAGTTAAACTCTTGGG[G/T]TTTTTTTTTCTTTTTTTTTGGCCAG
Phenotype
MIM: 614030
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPDYC PubMed Links

Gene Details

Gene
SPDYC
Gene Name
speedy/RINGO cell cycle regulator family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008778.1 Intron NP_001008778.1
XM_017017705.1 Intron XP_016873194.1
XM_017017706.1 Intron XP_016873195.1
XM_017017707.1 Intron XP_016873196.1
XM_017017708.1 Intron XP_016873197.1
XM_017017709.1 Intron XP_016873198.1
XM_017017710.1 Intron XP_016873199.1

View Full Product Details