Product Details

SNP ID
rs12223658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66012490 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAATTAAACCCTGTATTGCACTA[C/T]CTGCCTTCCTGGCTTGCCTGGTCCT
Phenotype
MIM: 603811 MIM: 606389 MIM: 601891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BANF1 PubMed Links

Gene Details

Gene
BANF1
Gene Name
barrier to autointegration factor 1
There are no transcripts associated with this gene.

Gene
CATSPER1
Gene Name
cation channel sperm associated 1
There are no transcripts associated with this gene.

Gene
CST6
Gene Name
cystatin E/M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323.3 Intron NP_001314.1

View Full Product Details