Product Details

SNP ID
rs7925040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66621825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATTCCTTATTCTGTAGCTGTTC[A/G]TATCTTACAAGGCACTTTTGTCTTT
Phenotype
MIM: 612409
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM14 PubMed Links

Gene Details

Gene
RBM14
Gene Name
RNA binding motif protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198836.1 Intron NP_001185765.1
NM_001198837.1 Intron NP_001185766.1
NM_006328.3 Intron NP_006319.1
Gene
RBM14-RBM4
Gene Name
RBM14-RBM4 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198845.1 Intron NP_001185774.1
NM_001198846.1 Intron NP_001185775.1

View Full Product Details