Product Details

SNP ID
rs17496724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55554623 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGTAACCATTCTCAGCAGCCCTG[A/C]TCTTCTGGTGTCTCCTATGTACTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR4C15 PubMed Links

Gene Details

Gene
OR4C15
Gene Name
olfactory receptor family 4 subfamily C member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001920.1 317 Missense Mutation GAT,GCT D106A NP_001001920.1

View Full Product Details