Product Details

SNP ID
rs11554863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61964817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCGCTCCCATCTTGCGCAAGTT[G/T]GTCACGTGGTCACCCAATTCTTTGA
Phenotype
MIM: 607854 MIM: 134770
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BEST1 PubMed Links

Gene Details

Gene
BEST1
Gene Name
bestrophin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139443.1 697 Intron NP_001132915.1
NM_001300786.1 697 Intron NP_001287715.1
NM_001300787.1 697 Intron NP_001287716.1
NM_004183.3 697 Intron NP_004174.1
XM_005274210.3 697 Intron XP_005274267.1
XM_005274215.3 697 Intron XP_005274272.1
XM_005274216.3 697 Intron XP_005274273.1
XM_005274219.3 697 Intron XP_005274276.1
XM_005274221.3 697 Intron XP_005274278.1
XM_011545229.2 697 Intron XP_011543531.1
XM_011545230.2 697 Intron XP_011543532.1
XM_011545233.2 697 Intron XP_011543535.1
XM_017018228.1 697 Intron XP_016873717.1
XM_017018229.1 697 Intron XP_016873718.1
XM_017018230.1 697 Intron XP_016873719.1
Gene
FTH1
Gene Name
ferritin heavy chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002032.2 697 Silent Mutation ACA,ACC T154T NP_002023.2
Gene
LOC399900
Gene Name
uncharacterized LOC399900
There are no transcripts associated with this gene.

View Full Product Details