Product Details

SNP ID
rs7129637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62271280 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTACCCAGAGAATCCTCAGTGGA[A/T]GATAAATGAATAGGGCAAGAGAGGA
Phenotype
MIM: 605562
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SCGB2A2 PubMed Links

Gene Details

Gene
SCGB2A2
Gene Name
secretoglobin family 2A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002411.3 Intron NP_002402.1
XM_005274005.3 Intron XP_005274062.3

View Full Product Details