Product Details
- SNP ID
-
rs6592380
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:71834789 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCTTAGTGCCATCCCTATCATCTG[C/T]CTCACTCCATCACTGGGTGATCTTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB108B
PubMed Links
Gene Details
- Gene
- DEFB108B
- Gene Name
- defensin beta 108B
View Full Product Details