Product Details

SNP ID
rs5743659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:71998832 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGCCTGCCCTTCCCTCCTGTTAG[A/G]GTTGGCTCTCGAGCTTGTGTGCCAG
Phenotype
MIM: 604113 MIM: 164009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL18BP PubMed Links

Gene Details

Gene
IL18BP
Gene Name
interleukin 18 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039659.1 889 Intron NP_001034748.1
NM_001039660.1 889 Intron NP_001034749.1
NM_001145055.1 889 Intron NP_001138527.1
NM_001145057.1 889 Intron NP_001138529.1
NM_005699.3 889 Intron NP_005690.2
NM_173042.2 889 Intron NP_766630.2
NM_173044.2 889 Intron NP_766632.2
XM_017017059.1 889 Intron XP_016872548.1
XM_017017060.1 889 Intron XP_016872549.1
XM_017017061.1 889 Intron XP_016872550.1
XM_017017062.1 889 Intron XP_016872551.1
XM_017017063.1 889 UTR 5 XP_016872552.1
Gene
NUMA1
Gene Name
nuclear mitotic apparatus protein 1
There are no transcripts associated with this gene.

Gene
RNF121
Gene Name
ring finger protein 121
There are no transcripts associated with this gene.

View Full Product Details