Product Details

SNP ID
rs7929180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77880557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGATGAATCTCCCACTTAGGG[A/T]GGAATCAAAATAGGTACCTATATCT
Phenotype
MIM: 611348 MIM: 608522
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AAMDC PubMed Links

Gene Details

Gene
AAMDC
Gene Name
adipogenesis associated Mth938 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316957.1 Intron NP_001303886.1
NM_001316958.1 Intron NP_001303887.1
NM_001316960.1 Intron NP_001303889.1
NM_001316961.1 Intron NP_001303890.1
NM_001316962.1 Intron NP_001303891.1
NM_024684.3 Intron NP_078960.1
XM_011544968.2 Intron XP_011543270.1
XM_011544969.2 Intron XP_011543271.1
XM_017017620.1 Intron XP_016873109.1
Gene
INTS4
Gene Name
integrator complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033547.3 Intron NP_291025.3
XM_011545352.2 Intron XP_011543654.1
XM_011545353.2 Intron XP_011543655.1
XM_017018560.1 Intron XP_016874049.1
Gene
RSF1
Gene Name
remodeling and spacing factor 1
There are no transcripts associated with this gene.

View Full Product Details