Product Details
- SNP ID
-
rs6895604
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:135534286 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAGGCAAGAAACTTGGCGACTTAT[A/T]TTTAGTTTTTATTTACATGAAGCGT
- Phenotype
-
MIM: 601726
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NEUROG1
PubMed Links
Gene Details
- Gene
- NEUROG1
- Gene Name
- neurogenin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006161.2 |
1664 |
UTR 3 |
|
|
NP_006152.2 |
View Full Product Details