Product Details

SNP ID
rs4997600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125905585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTTTCATGATGAATAGAACATA[A/G]GTTTTACTAATGCTATAATCAGTAT
Phenotype
MIM: 607960 MIM: 611640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX32 PubMed Links

Gene Details

Gene
DHX32
Gene Name
DEAH-box helicase 32 (putative)
There are no transcripts associated with this gene.

Gene
FANK1
Gene Name
fibronectin type III and ankyrin repeat domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145235.3 Intron NP_660278.3
XM_005270280.3 Intron XP_005270337.1
XM_006718058.3 Intron XP_006718121.1
XM_006718064.3 Intron XP_006718127.1
XM_011540348.2 Intron XP_011538650.1
XM_011540349.2 Intron XP_011538651.1
XM_011540350.2 Intron XP_011538652.1
XM_011540351.2 Intron XP_011538653.1
XM_011540356.2 Intron XP_011538658.1
XM_011540357.2 Intron XP_011538659.1
XM_011540358.2 Intron XP_011538660.1
XM_011540360.2 Intron XP_011538662.1
XM_011540361.2 Intron XP_011538663.1
XM_011540362.2 Intron XP_011538664.1
XM_011540363.2 Intron XP_011538665.1
XM_017016916.1 Intron XP_016872405.1
XM_017016917.1 Intron XP_016872406.1
XM_017016918.1 Intron XP_016872407.1
XM_017016919.1 Intron XP_016872408.1
XM_017016920.1 Intron XP_016872409.1
XM_017016921.1 Intron XP_016872410.1
XM_017016922.1 Intron XP_016872411.1
XM_017016923.1 Intron XP_016872412.1
XM_017016924.1 Intron XP_016872413.1
XM_017016925.1 Intron XP_016872414.1
XM_017016926.1 Intron XP_016872415.1

View Full Product Details