Product Details
- SNP ID
-
rs7089970
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:124447550 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCGGACGGGAAGAGGACATTGGCC[G/T]CGGAGTAGGAGGGGAAAGTCTGGAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NKX1-2
PubMed Links
Gene Details
- Gene
- NKX1-2
- Gene Name
- NK1 homeobox 2
View Full Product Details