Product Details

SNP ID
rs7928305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:695842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGGTAAGATGGCGGCCCCGCG[A/G]CGAGGTGAGCTCGGGCGGGGTGGGG
Phenotype
MIM: 602635 MIM: 614988
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEAF1 PubMed Links

Gene Details

Gene
DEAF1
Gene Name
DEAF1, transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293634.1 252 Intron NP_001280563.1
NM_021008.3 252 Intron NP_066288.2
XM_011519842.2 252 Intron XP_011518144.1
Gene
EPS8L2
Gene Name
EPS8 like 2
There are no transcripts associated with this gene.

Gene
TMEM80
Gene Name
transmembrane protein 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042463.2 252 Silent Mutation CGA,CGG R78R NP_001035928.2
NM_001276253.1 252 Silent Mutation CGA,CGG R78R NP_001263182.1
NM_001276274.1 252 Silent Mutation CGA,CGG R78R NP_001263203.1
NM_174940.3 252 Intron NP_777600.3
XM_006718206.3 252 Silent Mutation CGA,CGG R78R XP_006718269.3
XM_017017600.1 252 Intron XP_016873089.1

View Full Product Details