Product Details

SNP ID
rs11546821
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2213839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGATTATTCTTCACATCCCCTAG[A/G]GTGGAGACTCCGGTGAGGGTCAGGA
Phenotype
MIM: 603022 MIM: 612190 MIM: 172280
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BRICD5 PubMed Links
Additional Information
For this assay, SNP(s) [rs116977380] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
BRICD5
Gene Name
BRICHOS domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182563.3 899 Intron NP_872369.2
XM_005255263.4 899 Intron XP_005255320.1
Gene
E4F1
Gene Name
E4F transcription factor 1
There are no transcripts associated with this gene.

Gene
MLST8
Gene Name
MTOR associated protein, LST8 homolog
There are no transcripts associated with this gene.

Gene
PGP
Gene Name
phosphoglycolate phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042371.2 899 Silent Mutation ACC,ACT T285T NP_001035830.1

View Full Product Details