Product Details

SNP ID
rs8045864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2494103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATAGGCTGTGACTTTCTTAGCAC[A/G]CACAGCTGGTTAAAGAGAACTGGGA
Phenotype
MIM: 613577
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBC1D24 PubMed Links

Gene Details

Gene
TBC1D24
Gene Name
TBC1 domain family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199107.1 Intron NP_001186036.1
NM_020705.2 Intron NP_065756.1
XM_017023493.1 Intron XP_016878982.1
XM_017023494.1 Intron XP_016878983.1
XM_017023495.1 Intron XP_016878984.1

View Full Product Details