Product Details
- SNP ID
-
rs4785983
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:5046752 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTTCCCTTTCTTCTACGTTATTT[A/C]TTGACTTTCTTCTACGTTATTTCTT
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C16orf89
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76713390] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C16orf89
- Gene Name
- chromosome 16 open reading frame 89
- Gene
- NAGPA-AS1
- Gene Name
- NAGPA antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details