Product Details
- SNP ID
-
rs4786664
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:5299134 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTGTTCTAACAATTTCCATCACAA[G/T]AAATTAAATTTGCCACTTCTTGGGC
- Phenotype
-
MIM: 605104
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RBFOX1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115067305] are located under a probe and SNP(s) [rs115883086] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RBFOX1
- Gene Name
- RNA binding protein, fox-1 homolog 1
View Full Product Details