Product Details

SNP ID
rs11557470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7682425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCAGTCGCGCCTGGCCGCGCTG[G/T]GCCGCCAGGTGGGCGCGCGCGTGCT
Phenotype
MIM: 151445 MIM: 609565
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FCER2 PubMed Links

Gene Details

Gene
FCER2
Gene Name
Fc fragment of IgE receptor II
There are no transcripts associated with this gene.

Gene
MCEMP1
Gene Name
mast cell expressed membrane protein 1
There are no transcripts associated with this gene.

Gene
TRAPPC5
Gene Name
trafficking protein particle complex 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042461.2 242 Missense Mutation GGC,TGC G58C NP_001035926.1
NM_001042462.1 242 Missense Mutation GGC,TGC G58C NP_001035927.1
NM_174894.2 242 Missense Mutation GGC,TGC G58C NP_777554.1

View Full Product Details