Product Details

SNP ID
rs13302936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8867841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGCCAGCGGGAAGGGCAGGGTG[G/T]CCGGGGGCCCAGCGGCGGCTGCGGC
Phenotype
MIM: 142992
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HMX1 PubMed Links

Gene Details

Gene
HMX1
Gene Name
H6 family homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306142.1 1102 Intron NP_001293071.1
NM_018942.2 1102 Missense Mutation GAC,GCC D300A NP_061815.2

View Full Product Details