Product Details

SNP ID
rs9474379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:53062092 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAACCATCAAGACTTAGAAATGGA[C/G]GCTGTCGATGGACGTTCTGAGCACA
Phenotype
MIM: 609091 MIM: 612325
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXO9 PubMed Links

Gene Details

Gene
FBXO9
Gene Name
F-box protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012347.4 Intron NP_036479.1
NM_033480.2 Intron NP_258441.1
NM_033481.3 Intron NP_258442.2
XM_005248995.4 Intron XP_005249052.1
XM_005248996.4 Intron XP_005249053.1
Gene
ICK
Gene Name
intestinal cell (MAK-like) kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014920.3 Intron NP_055735.1
NM_016513.4 Intron NP_057597.2
XM_011514419.1 Intron XP_011512721.1
XM_011514420.2 Intron XP_011512722.1
XM_011514421.1 Intron XP_011512723.1
XM_017010485.1 Intron XP_016865974.1
XM_017010486.1 Intron XP_016865975.1
XM_017010487.1 Intron XP_016865976.1
XM_017010488.1 Intron XP_016865977.1
XM_017010489.1 Intron XP_016865978.1
XM_017010490.1 Intron XP_016865979.1
XM_017010491.1 Intron XP_016865980.1
XM_017010492.1 Intron XP_016865981.1
Gene
LOC105375092
Gene Name
uncharacterized LOC105375092
There are no transcripts associated with this gene.

View Full Product Details