Product Details
- SNP ID
-
rs12936979
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:20869021 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTGTGAAAAGACAGTGGGGAGAA[A/C]ATATAGTGAGGGGGAAAAGTTTTTG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC144NL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7209605] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC144NL
- Gene Name
- coiled-coil domain containing 144 family, N-terminal like
- Gene
- CCDC144NL-AS1
- Gene Name
- CCDC144NL antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details