Product Details

SNP ID
rs610142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15678120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTATTTCCATCCTGTTTACCTTCA[A/T]ATGCTAATCTCTTCTCAAACACCCT
Phenotype
MIM: 611485
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP4F12 PubMed Links

Gene Details

Gene
CYP4F12
Gene Name
cytochrome P450 family 4 subfamily F member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023944.3 Intron NP_076433.3
XM_006722850.3 Intron XP_006722913.1
XM_011528202.2 Intron XP_011526504.1
XM_011528203.2 Intron XP_011526505.1
XM_011528204.2 Intron XP_011526506.1
XM_011528205.2 Intron XP_011526507.1
XM_011528207.2 Intron XP_011526509.1
XM_011528208.2 Intron XP_011526510.1
XM_017027172.1 Intron XP_016882661.1

View Full Product Details