Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286587.1 | Intron | NP_001273516.1 | |||
NM_001286588.1 | Intron | NP_001273517.1 | |||
NM_001286589.1 | Intron | NP_001273518.1 | |||
NM_016108.3 | Intron | NP_057192.2 | |||
XM_011535881.2 | Intron | XP_011534183.1 | |||
XM_011535882.2 | Intron | XP_011534184.1 | |||
XM_011535883.2 | Intron | XP_011534185.1 | |||
XM_011535884.2 | Intron | XP_011534186.1 | |||
XM_011535885.2 | Intron | XP_011534187.1 | |||
XM_017010927.1 | Intron | XP_016866416.1 | |||
XM_017010928.1 | Intron | XP_016866417.1 |