Product Details

SNP ID
rs2860194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103477030 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCTTCTTAAATCCTCAGTGGGA[A/C]AATTCTGAGATGTGTTCTATGCATG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CALHM3 PubMed Links

Gene Details

Gene
CALHM3
Gene Name
calcium homeostasis modulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129742.1 Intron NP_001123214.1
Gene
NEURL1-AS1
Gene Name
NEURL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details