Product Details

SNP ID
rs28649027
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:1724489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGACTCCAAAAGAAACTGCGCC[C/G]TGAGCTGCACTTCCGACTTCGGCGC
Phenotype
MIM: 116951
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDK11A PubMed Links

Gene Details

Gene
CDK11A
Gene Name
cyclin dependent kinase 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313896.1 77 UTR 5 NP_001300825.1
NM_001313982.1 77 UTR 5 NP_001300911.1
NM_024011.3 77 UTR 5 NP_076916.2
NM_033529.3 77 UTR 5 NP_277071.2
Gene
SLC35E2
Gene Name
solute carrier family 35 member E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199787.1 77 Intron NP_001186716.1
NM_182838.2 77 Intron NP_878258.1

View Full Product Details