Product Details

SNP ID
rs1062090
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:1752461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGCCAGGGGCTTCCCTGCAGAA[A/G]TTTTAGGGGAAGAGGTGCAGGTCAA
Phenotype
MIM: 611616
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NADK PubMed Links

Gene Details

Gene
NADK
Gene Name
NAD kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198993.1 1710 UTR 3 NP_001185922.1
NM_001198994.1 1710 UTR 3 NP_001185923.1
NM_001198995.1 1710 UTR 3 NP_001185924.1
NM_023018.4 1710 UTR 3 NP_075394.3
XM_005244778.2 1710 UTR 3 XP_005244835.1
XM_006710837.2 1710 UTR 3 XP_006710900.1
XM_006710838.2 1710 UTR 3 XP_006710901.1
XM_006710839.1 1710 UTR 3 XP_006710902.1
XM_011542006.1 1710 UTR 3 XP_011540308.1
XM_011542007.1 1710 UTR 3 XP_011540309.1
XM_011542008.1 1710 UTR 3 XP_011540310.1
XM_011542009.2 1710 UTR 3 XP_011540311.1
XM_017002143.1 1710 UTR 3 XP_016857632.1
XM_017002144.1 1710 UTR 3 XP_016857633.1
XM_017002145.1 1710 UTR 3 XP_016857634.1
XM_017002146.1 1710 UTR 3 XP_016857635.1
Gene
SLC35E2
Gene Name
solute carrier family 35 member E2
There are no transcripts associated with this gene.

View Full Product Details