Product Details

SNP ID
rs4970429
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:975523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGACCCCCACCCTGCAGGGGC[C/T]GCTGCACTGGGGAGGGGCAGGCAGA
Phenotype
MIM: 615921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL17 PubMed Links

Gene Details

Gene
KLHL17
Gene Name
kelch like family member 17
There are no transcripts associated with this gene.

Gene
PERM1
Gene Name
PPARGC1 and ESRR induced regulator, muscle 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291366.1 2740 UTR 3 NP_001278295.1
NM_001291367.1 2740 UTR 3 NP_001278296.1
XM_017002583.1 2740 UTR 3 XP_016858072.1
XM_017002584.1 2740 UTR 3 XP_016858073.1
XM_017002585.1 2740 UTR 3 XP_016858074.1
Gene
PLEKHN1
Gene Name
pleckstrin homology domain containing N1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160184.1 2740 Intron NP_001153656.1
NM_032129.2 2740 Intron NP_115505.2
XM_006710944.3 2740 Intron XP_006711007.2
XM_011542248.2 2740 Intron XP_011540550.2
XM_017002474.1 2740 Intron XP_016857963.1
XM_017002475.1 2740 Intron XP_016857964.1
XM_017002476.1 2740 Intron XP_016857965.1
XM_017002477.1 2740 Intron XP_016857966.1
XM_017002478.1 2740 Intron XP_016857967.1
XM_017002479.1 2740 Intron XP_016857968.1

View Full Product Details