Product Details

SNP ID
rs35626575
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:1210896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGGGCCAGCTGCCCTTGTCAG[C/T]GCCTGGCCAGGAACCTCCACCCCAG
Phenotype
MIM: 614282 MIM: 603905 MIM: 600315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDF4 PubMed Links

Gene Details

Gene
SDF4
Gene Name
stromal cell derived factor 4
There are no transcripts associated with this gene.

Gene
TNFRSF18
Gene Name
TNF receptor superfamily member 18
There are no transcripts associated with this gene.

Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003327.3 Intron NP_003318.1
XM_011542074.2 Intron XP_011540376.1
XM_011542075.2 Intron XP_011540377.1
XM_011542076.2 Intron XP_011540378.1
XM_011542077.2 Intron XP_011540379.1
XM_017002231.1 Intron XP_016857720.1
XM_017002232.1 Intron XP_016857721.1

View Full Product Details