Product Details

SNP ID
rs34034855
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:161047603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGAGAAGCTGGAGGGCCAAAG[C/T]CTATAGAGTTGGGCACTACAGCTGC
Phenotype
MIM: 614264 MIM: 616041 MIM: 191523
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP30 PubMed Links

Gene Details

Gene
ARHGAP30
Gene Name
Rho GTPase activating protein 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025598.1 3233 UTR 3 NP_001020769.1
NM_001287600.1 3233 UTR 3 NP_001274529.1
NM_001287602.1 3233 UTR 3 NP_001274531.1
NM_181720.2 3233 UTR 3 NP_859071.2
XM_005245070.2 3233 UTR 3 XP_005245127.1
XM_005245073.3 3233 UTR 3 XP_005245130.1
XM_011509391.2 3233 UTR 3 XP_011507693.1
XM_017000960.1 3233 UTR 3 XP_016856449.1
Gene
TSTD1
Gene Name
thiosulfate sulfurtransferase like domain containing 1
There are no transcripts associated with this gene.

Gene
USF1
Gene Name
upstream transcription factor 1
There are no transcripts associated with this gene.

View Full Product Details