Product Details
- SNP ID
-
rs28377275
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
37
- Location
-
Chr.1:170946357 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACCAGTGCTCTTCAAAACTGTCA[A/G]GGTCATAAACAAGGAAAGACTGCTA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MROH9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78346161] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MROH9
- Gene Name
- maestro heat like repeat family member 9
View Full Product Details