Product Details

SNP ID
rs28384819
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:171249786 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCATAAATAGAGGGCTGGATACAA[A/C]TCTGACAGTGCCAATGGCTTCCTAG
Phenotype
MIM: 136130
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FMO1 PubMed Links

Gene Details

Gene
FMO1
Gene Name
flavin containing monooxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282692.1 Intron NP_001269621.1
NM_001282693.1 Intron NP_001269622.1
NM_001282694.1 Intron NP_001269623.1
NM_002021.2 Intron NP_002012.1
XM_005245037.3 Intron XP_005245094.1
XM_005245038.3 Intron XP_005245095.1
XM_006711241.3 Intron XP_006711304.1
XM_006711242.3 Intron XP_006711305.1

View Full Product Details