Product Details
- SNP ID
-
rs28616098
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:182447970 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCAGACCTAGAAGCTCTGAAATTT[A/T]AAAAAAAAAAAAAAAGCATTTTACC
- Phenotype
-
MIM: 611012
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RGSL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs558278159] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RGSL1
- Gene Name
- regulator of G-protein signaling like 1
View Full Product Details