Product Details

SNP ID
rs28924726
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:153002687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTAGAGGATTCTTCAAAGAGTGT[C/G]TAAGCGAAAGCTTCAGAGTATATAT
Phenotype
MIM: 182271
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPRR3 PubMed Links

Gene Details

Gene
SPRR3
Gene Name
small proline rich protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097589.1 Intron NP_001091058.1
NM_005416.2 Intron NP_005407.1

View Full Product Details