Product Details

SNP ID
rs35923929
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:193101127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAGGAAAGTTTTTCAATCATCT[C/G]CCACATCACTCACTTGGATCTGGTT
Phenotype
MIM: 606820 MIM: 600063
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLRX2 PubMed Links

Gene Details

Gene
GLRX2
Gene Name
glutaredoxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243399.1 Intron NP_001230328.1
NM_001319291.1 Intron NP_001306220.1
NM_016066.4 Intron NP_057150.2
NM_197962.2 Intron NP_932066.1
XM_017001397.1 Intron XP_016856886.1
Gene
TROVE2
Gene Name
TROVE domain family member 2
There are no transcripts associated with this gene.

View Full Product Details