Product Details

SNP ID
rs28679436
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:154323968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCCCTTTCTCCCTTGCTTCCC[C/T]CCCAACTTTTCACTGAAACAGTAAG
Phenotype
MIM: 606578 MIM: 605867
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AQP10 PubMed Links

Gene Details

Gene
AQP10
Gene Name
aquaporin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080429.2 Intron NP_536354.2
XM_011510104.2 Intron XP_011508406.1
Gene
ATP8B2
Gene Name
ATPase phospholipid transporting 8B2
There are no transcripts associated with this gene.

View Full Product Details