Product Details

SNP ID
hCV57591956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:155219013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTTACTCCTGCAAGTAATTCTG[A/G]CTTCCCAATGTGGATGGGTCATGTG
Phenotype
MIM: 600605 MIM: 188062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBAP1 PubMed Links

Gene Details

Gene
GBAP1
Gene Name
glucosylceramidase beta pseudogene 1
There are no transcripts associated with this gene.

Gene
MTX1
Gene Name
metaxin 1
There are no transcripts associated with this gene.

Gene
THBS3
Gene Name
thrombospondin 3
There are no transcripts associated with this gene.

View Full Product Details