Product Details

SNP ID
rs28539337
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:156669407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACCCTGGCCCAGGTGAACAGGA[G/T]CCCCTGCCCAAGAGGTCTTCAGAGC
Phenotype
MIM: 600347 MIM: 600915
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCAN PubMed Links

Gene Details

Gene
BCAN
Gene Name
brevican
There are no transcripts associated with this gene.

Gene
NES
Gene Name
nestin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006617.1 4914 Missense Mutation GAT,GCT D1594A NP_006608.1

View Full Product Details