Product Details
- SNP ID
-
rs28623134
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:16975445 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATAGCCCAGACAGAACCTGGCAC[G/T]GGAGCCCGGACAGAACCTGGCACGG
- Phenotype
-
MIM: 615776
MIM: 156790
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CROCC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3830850] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CROCC
- Gene Name
- ciliary rootlet coiled-coil, rootletin
There are no transcripts associated with this gene.
- Gene
- MFAP2
- Gene Name
- microfibrillar associated protein 2
View Full Product Details