Product Details

SNP ID
rs28411938
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:17410908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGATAGGCCCCAGAAAACATCCT[C/T]CCTCCTCTAAGAACTATCTGTAGAA
Phenotype
MIM: 610363 MIM: 609587
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PADI6 PubMed Links

Gene Details

Gene
PADI6
Gene Name
peptidyl arginine deiminase 6
There are no transcripts associated with this gene.

Gene
RCC2
Gene Name
regulator of chromosome condensation 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136204.2 Intron NP_001129676.1
NM_018715.3 Intron NP_061185.1

View Full Product Details