Product Details

SNP ID
rs866489567
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:23845098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTTTTTTTGGTAGAAATGATTG[G/T]GATGTACAAATTTTTTATTTGATCA
Phenotype
MIM: 612280
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FUCA1 PubMed Links

Gene Details

Gene
FUCA1
Gene Name
fucosidase, alpha-L- 1, tissue
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000147.4 1686 UTR 3 NP_000138.2
XM_005245821.2 1686 UTR 3 XP_005245878.1
XM_011541167.2 1686 UTR 3 XP_011539469.1
XM_017000905.1 1686 UTR 3 XP_016856394.1

View Full Product Details