Product Details

SNP ID
rs12074469
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:25889837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGTATGGGGTCCCCTTGCCCA[C/T]CTCATCCAGCCAAGAGGGCCTCACT
Phenotype
MIM: 151442
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STMN1 PubMed Links

Gene Details

Gene
STMN1
Gene Name
stathmin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145454.2 Intron NP_001138926.1
NM_005563.3 Intron NP_005554.1
NM_203399.1 Intron NP_981944.1
NM_203401.1 Intron NP_981946.1

View Full Product Details