Product Details

SNP ID
rs28435104
Assay Type
Functionally Tested
NCBI dbSNP Submissions
7
Location
Chr.1:235370738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTTTATGCTTTTTTCCTCCTT[G/T]TCTTTTCTTTTTTTTTTTTTTGAGA
Phenotype
MIM: 604934
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
TBCE PubMed Links

Gene Details

Gene
TBCE
Gene Name
tubulin folding cofactor E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079515.2 Intron NP_001072983.1
NM_001287801.1 Intron NP_001274730.1
NM_001287802.1 Intron NP_001274731.1
NM_003193.4 Intron NP_003184.1

View Full Product Details