Product Details

SNP ID
rs28465248
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:241997767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTGGGGGATGCTTTTCTCTCTC[A/T]GTGTTCTGAGCAGGGAACGTTTCCT
Phenotype
MIM: 609605
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MAP1LC3C PubMed Links

Gene Details

Gene
MAP1LC3C
Gene Name
microtubule associated protein 1 light chain 3 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004343.2 Intron NP_001004343.1
XM_005273139.3 Intron XP_005273196.1

View Full Product Details