Product Details
- SNP ID
-
rs8661
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
25
- Location
-
Chr.1:16605556 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAACTGGTGACACACAGAACAACA[A/T]GGATGAATCTCAAAAACATTTGGAG
- Phenotype
-
MIM: 610501
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NBPF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs575865507] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NBPF1
- Gene Name
- neuroblastoma breakpoint family member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017940.4 |
|
Intron |
|
|
NP_060410.2 |
View Full Product Details