Product Details

SNP ID
rs28900411
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233775538 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCTCCCAAGATCCCACAAGAGAC[A/T]GACTGGATTCTCTCTGGCCTGGAGT
Phenotype
MIM: 191740 MIM: 606435 MIM: 606428 MIM: 606429 MIM: 606430 MIM: 606431 MIM: 606432 MIM: 606433 MIM: 606434
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
MROH2A PubMed Links
Additional Information
For this assay, SNP(s) [rs115692048] are located under a probe and SNP(s) [rs114112305] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MROH2A
Gene Name
maestro heat like repeat family member 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287395.1 1517 Intron NP_001274324.1
XM_011511075.2 1517 Intron XP_011509377.1
XM_011511076.2 1517 Intron XP_011509378.1
XM_011511077.2 1517 Intron XP_011509379.1
XM_011511078.2 1517 Intron XP_011509380.1
XM_011511079.2 1517 Intron XP_011509381.1
XM_011511080.2 1517 UTR 5 XP_011509382.1
XM_011511081.2 1517 Intron XP_011509383.1
XM_011511082.2 1517 Intron XP_011509384.1
XM_011511083.2 1517 Intron XP_011509385.1
XM_011511084.2 1517 Intron XP_011509386.1
XM_011511085.2 1517 Intron XP_011509387.1
XM_011511086.2 1517 Intron XP_011509388.1
Gene
UGT1A1
Gene Name
UDP glucuronosyltransferase family 1 member A1
There are no transcripts associated with this gene.

Gene
UGT1A10
Gene Name
UDP glucuronosyltransferase family 1 member A10
There are no transcripts associated with this gene.

Gene
UGT1A3
Gene Name
UDP glucuronosyltransferase family 1 member A3
There are no transcripts associated with this gene.

Gene
UGT1A4
Gene Name
UDP glucuronosyltransferase family 1 member A4
There are no transcripts associated with this gene.

Gene
UGT1A5
Gene Name
UDP glucuronosyltransferase family 1 member A5
There are no transcripts associated with this gene.

Gene
UGT1A6
Gene Name
UDP glucuronosyltransferase family 1 member A6
There are no transcripts associated with this gene.

Gene
UGT1A7
Gene Name
UDP glucuronosyltransferase family 1 member A7
There are no transcripts associated with this gene.

Gene
UGT1A8
Gene Name
UDP glucuronosyltransferase family 1 member A8
There are no transcripts associated with this gene.

Gene
UGT1A9
Gene Name
UDP glucuronosyltransferase family 1 member A9
There are no transcripts associated with this gene.

View Full Product Details