Product Details

SNP ID
hCV57749080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10124810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAACATACATTCTGAAATGTAT[A/G]GTCTTCTTATTGACACTTACATAAA
Phenotype
MIM: 180390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RRM2 PubMed Links

Gene Details

Gene
RRM2
Gene Name
ribonucleotide reductase regulatory subunit M2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034.3 592 Missense Mutation AGT,GGT S177G NP_001025.1
NM_001165931.1 592 Missense Mutation AGT,GGT S237G NP_001159403.1

View Full Product Details