Product Details

SNP ID
rs28365982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:159712509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCACTTAACGGTGGTGGCTGGT[G/T]CTGCGCCGGATCCGGGAGAGGGGCG
Phenotype
MIM: 605683 MIM: 613334
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BAZ2B PubMed Links

Gene Details

Gene
BAZ2B
Gene Name
bromodomain adjacent to zinc finger domain 2B
There are no transcripts associated with this gene.

Gene
MARCH7
Gene Name
membrane associated ring-CH-type finger 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282805.1 21 UTR 5 NP_001269734.1
NM_001282806.1 21 UTR 5 NP_001269735.1
NM_001282807.1 21 Intron NP_001269736.1
NM_022826.3 21 UTR 5 NP_073737.1
XM_005246773.2 21 UTR 5 XP_005246830.1
XM_005246774.1 21 UTR 5 XP_005246831.1
XM_017004718.1 21 UTR 5 XP_016860207.1
XM_017004719.1 21 UTR 5 XP_016860208.1
XM_017004720.1 21 UTR 5 XP_016860209.1
XM_017004721.1 21 UTR 5 XP_016860210.1

View Full Product Details