Product Details

SNP ID
rs28375809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:161312174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTTTTTGAGATGGAGTAT[C/T]GTTCTGTCACCCAGGCTGGAGTGCG
Phenotype
MIM: 607173
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMD14 PubMed Links

Gene Details

Gene
PSMD14
Gene Name
proteasome 26S subunit, non-ATPase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005805.5 Intron NP_005796.1

View Full Product Details