Product Details

SNP ID
rs35392215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201235371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGTATTCTGGAAGGTACTTTT[C/G]TTAAAATTACAGTATTTAGTCCATT
Phenotype
MIM: 601762 MIM: 601763
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CASP10 PubMed Links

Gene Details

Gene
CASP10
Gene Name
caspase 10
There are no transcripts associated with this gene.

Gene
CASP8
Gene Name
caspase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080124.1 Intron NP_001073593.1
NM_001080125.1 Intron NP_001073594.1
NM_001228.4 Intron NP_001219.2
NM_033355.3 Intron NP_203519.1
NM_033356.3 Intron NP_203520.1
NM_033358.3 Intron NP_203522.1
XM_005246885.1 Intron XP_005246942.1
XM_005246886.1 Intron XP_005246943.1
XM_005246887.1 Intron XP_005246944.1
XM_005246888.1 Intron XP_005246945.1
XM_005246889.1 Intron XP_005246946.1
XM_005246890.3 Intron XP_005246947.1
XM_005246891.4 Intron XP_005246948.1
XM_005246892.1 Intron XP_005246949.1
XM_005246893.2 Intron XP_005246950.1
XM_005246894.3 Intron XP_005246951.1
XM_005246895.2 Intron XP_005246952.1
XM_006712789.1 Intron XP_006712852.1
XM_006712790.3 Intron XP_006712853.1
XM_006712793.2 Intron XP_006712856.1
XM_011511969.1 Intron XP_011510271.1

View Full Product Details